Learn More
Cole-Parmer Traceable Thermometer with Clock, Humidity Monitor, and Calibration
Supplier: Cole-Parmer 9008006
Unique unit simultaneously displays time-of-day, temperature, and humidity. Clock displays AM/PM or 24-hour military time to the exact minute with 0.01% accuracy. Noting changes is easy with triple display. Internal sensors make it ideal for use in hoods, storerooms, clean rooms, incubators, drying chambers, and environmental cabinets. Temperature range is 32.0 to 122.0F and 0.0 to 50.0C. Resolution is 0.1 and accuracy is 1C. Relative humidity range is 20 to 90%. Resolution is 1% RH and accuracy is 5% RH mid-range, otherwise 8%. At the touch of a button, memory recalls highest and lowest temperature and humidity readings.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.