Learn More
Cole-Parmer Environmental Express UC475-GN Ultimate Cup, Digestion Cups with Green Caps, 50 mL; 500/Pk
Supplier: Cole-Parmer 5320313
Approved for hot plate digestions and certified for low metals background.