Learn More
Chemglass Life Sciences Temperature Controller, Type "J", Supplied Complete with CG-15001-25 Thermocouple
Supplier: Chemglass Life Sciences CG1500130
Temperature Controller, Type "J", Supplied Complete with CG-15001-25 Thermocouple. PID controller for use with our CG-1100 oil baths. Controller has 1500 Watts of power and will control temperatures to ± 1.0°C. Supplied complete with CG-15001-25 Type "J" Thermocouple.