Learn More
Rice Lake Weighing System Scale,Floor Level 150-10-7 550lb(250kg) x 0.2lb(100g) includes 6 AA Alkaline batteries
Supplier: Rice Lake Weighing System 121304
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Medical Scale, Rice Lake Weighing Systems, Floor Level Physician Scale 150-107 550lb(250kg) x 0.2lb(100g) includes 6 AA batteries, LCD display, 14.5in x 14.5in x 3 in H, BMI (body mass index) function, Motion Trap