Learn More
Rice Lake Weighing System Scale, Hanging OS-25 25kg x 0.02kg/55lb x 0.05lb/55lb x 1/2oz Includes carrying case, 2 AA size Alkaline Batteries, manual Bottom hook fully attached, top hook removable
Supplier: Rice Lake Weighing System 130293
Medical Scale, Rice Lake Weighing Systems, Hanging OS-25 25kg x 0.02kg/55lb x 0.05lb/55lb x 1/2oz Includes carrying case, (2) AA Batteries, bottom hook fully attached, top hook removable, adjustable auto shutoff, units kg/lb/lb-oz