Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: TT (Tetrathionate) Broth Base, Hajna
Selective enrichment for Salmonella from food and dairy products
Supplier: BD 249110
Description
Specifications
ISO, 9000, USDA | |
Dehydrated | |
Microbiology Media | |
Isolation of Escherichia coli 25922, Salmonella enterica subspecies enterica serotype Typhimurium 14028 | |
2000 g |
<30°C | |
TT Broth Base, Hajna | |
Yeast extract 2g/L, Tryptose 18g/L, Dextrose 0.5g/L, D-Mannitol 2.5g/L, Sodium Desoxycholate 0.5g/L, Sodium Chloride 5g/L, Sodium Thiosulfate 38g/L, Calcium Carbonate 25g/L, Brilliant green 0.01g/L | |
Pail |
Safety and Handling
ShelfLife : 1825 Days
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction