Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Diethyl dimethylaminomethylenemalonate, 94%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
Description
Derivatives of this compound, diethyl ethoxymethylenemalonate is used in the synthesis of chemotherapeutic agents using 6-aminobenzothiazoles as starting material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Derivatives of this compound, diethyl ethoxymethylenemalonate is used in the synthesis of chemotherapeutic agents using 6-aminobenzothiazoles as starting material.
Solubility
Soluble in methanol.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Derivatives of this compound, diethyl ethoxymethylenemalonate is used in the synthesis of chemotherapeutic agents using 6-aminobenzothiazoles as starting material.
Solubility
Soluble in methanol.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Specifications
Boiling Point | 134°C to 136°C (0.5 mmHg) |
Quantity | 25 g |
Solubility Information | Soluble in methanol. |
Formula Weight | 215.25 |
Percent Purity | 95% |
Chemical Name or Material | Diethyl dimethylaminomethylenemalonate |
Safety and Handling
TSCA | No |
Recommended Storage | Keep cold |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction