Learn More
International Crystal 7 mm Die Set
Supplier: International Crystal 189
Die Set only. Hand press sold separate. The Quick Press is a hand press for making transparent KBr pellets (discs) with diameters of 7mm, 4mm or 2mm.The Quick Press uses the principal of toggle action to apply very high pressure with minimal force. The KBr pellet is formed using only the pressure of a firm handshake applied steadily to the handle.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.