Learn More
Electron Microscopy Sciences Diamond Wire
SDP
Supplier:  Electron Microscopy Sciences 7200910
Encompass_Preferred
SuperLok diamond wire has core wire with a tensile strength in excess of 400,000 lbs. An electro copper sheath with impregnated 20 to 120 micron diamonds is added. Guaranteed consistency in diamond impregnation. Wire is 6".
Catalog No. 50-949-242