Learn More
Electron Microscopy Sciences Diamond Stone, Medium
Supplier: Electron Microscopy Sciences 6208251
This patented process of heat treatment which bonds the highest quality industrial grade diamond particles in a stainless alloy to a precision matched metal substrate, has been imitated, but has never come close to being duplicated. Our stone which is set on a walnut pedestal with a cover. Medium grit (270), size: 2"x6" (51mmx152mm)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.