Learn More
Electron Microscopy Sciences Flexible Shaft Machine Diamond Points Tapered Wheel, 7.3 mm Tip, 073 I.S.O.
Supplier: Electron Microscopy Sciences 6215573
All diamond points are mounted on 3/32" (2.35 mm) shanks. Suitable for Flex Shaft machines. Exceptionally long lasting. Use at 10,000 RPM.
Tip: 7.3 mm
I.S.O. size: 073