Learn More
Diamedix™ Immunosimplicity™ Toxoplasma IgG Test Kit
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Detects IgG antibodies to Toxoplasma gondii in human serum
Supplier: Diamedix™ 720300
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Alert:
Handle samples, standards, controls and the materials that contact them as potential biohazards.
Certain reagents contain the preservatives ProClin™ 300 or Sodium Azide.
Includes: Antigen Wells, Standards: 0 IU/mL, 50 IU/mL, 250 IU/mL, High Positive Control, Low Positive Control, Negative Control, Sample A Diluent, Wash S Concentrate (20X), Conjugate, Substrate HRP, Stop M Solution
Description
The Diamedix™ Immunosimplicity™ Toxoplasma IgG Test Kit is a qualitative and quantitative indirect enzyme immunoassay
- Detects IgG antibodies to Toxoplasma gondii in human serum
- Aids in the assessment of a patient's immunological response to infection with Toxoplasma gondii
- Determines the immune status of individuals, including females of child-bearing age
- This product is not FDA cleared for use in screening blood and plasma donors
- For in vitro diagnostic use
- Reagent volumes/configurations are suitable for manual use, automated Mago™ systems, and Dynex DSX™ and DS2™ systems
Specifications
T. gondii IgG | |
Immunosimplicity™ Toxoplasma IgG Test Kit | |
Antigen wells, standards: 0 IU/mL, 50 IU/mL, 250 IU/mL, high positive control, low positive control, negative control, sample A diluent, wash S concentrate 20X, conjugate, substrate HRP, stop M solution | |
96 Tests |