Learn More
Diamedix™ Immunosimplicity™ HSV 1 and 2 IgM Test Kit
Detects IgM antibodies to herpes simplex virus type 1 and/or type 2 in human serum
Supplier: Diamedix™ 720350
Alert:
Handle samples, calibrators, controls and the materials that contact them as potential biohazards.
Certain reagents contain the preservatives ProClin™ 300 or Sodium Azide.
Includes: Antigen Wells, Cut-off Calibrator, Low Positive Control, Negative Control, Sample D Diluent, Wash S Concentrate (20X), Conjugate, Substrate G, Stop M Solution
Description
The Diamedix™ Immunosimplicity™ HSV 1 and 2 IgM Test Kit is a qualitative indirect enzyme immunoassay
- Detects IgM antibodies to herpes simplex virus (HSV) type 1 and/or type 2 in human serum
- Aids in the diagnosis of a primary or reactivated infection with HSV
- The performance of this assay has not been established for use in neonates, infants, or on cord blood and immunocompromised patients
- For in vitro diagnostic use
- Reagent volumes/configurations are suitable for manual use, automated Mago™ systems, and Dynex DSX™ and DS2™ systems
Specifications
IgM Antibodies | |
HSV 1 and 2 IGM ELISA Test Kit | |
Antigen wells, positive control, negative control, sample A diluent, wash S concentrate 20X, conjugate, substrate HRP, stop M solution | |
96 Tests |