Learn More
Diamedix™ Immunosimplicity™ HSV 1 and 2 IgG Test Kit
Detects IgG antibodies to herpes simplex virus type 1 and/or type 2 in human serum
Supplier: Diamedix™ 720340
Alert:
Handle samples, calibrator, controls and the materials that contact them as potential biohazards.
Certain reagents contain the preservatives ProClin™ 300 or Sodium Azide.
Includes: Antigen Wells, Positive Control, Negative Control, Sample A Diluent, Wash S Concentrate (20X), Conjugate, Substrate HRP, Stop M Solution
Description
The Diamedix™ Immunosimplicity™ HSV 1 and 2 IgG Test Kit is a qualitative and semi-quantitative indirect enzyme immunoassay
- Detects IgG antibodies to herpes simplex virus (HSV) type 1 and/or type 2 in human serum
- This test is useful for indicating a past infection with HSV in a single specimen, including females of child-bearing age
- The evaluation of acute and convalescent specimens, by demonstrating seroconversion or a significant increase in antibody level, can aid in the diagnosis of primary infection with HSV
- For in vitro diagnostic use
- Reagent volumes/configurations are suitable for manual use, automated Mago™ systems, and Dynex DSX™ and DS2™ systems
Specifications
IgG Antibodies | |
HSV 1 and 2 IGG ELISA Test Kit | |
Antigen wells, positive control, negative control, sample A diluent, wash S concentrate 20X, conjugate, substrate HRP, stop M solution | |
96 Tests |