Learn More
Diaion™ HP20, synthetic adsorbent resin, highly porous type, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 046488A1
Description
Diaion™ HP20 is used as a filler in chromatography columns due to its extremely large pores. It is considered as an effective adsorbent to eliminate toxic effects in fermentation and separation of traces heavy metal ions. It is also useful for the purification of peptides, proteins, fermentation products, polyphenols and cephalosporin C. It helps to remove bitter tasting substances from fruit juices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Beads | |
Highly porous type | |
1.01 g/mL |
Diaion™ HP20, synthetic adsorbent resin | |
≥250μm | |
1 kg |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only