Learn More
Strem, An Ascensus Company CAS# 683-18-1. 50g. Di-n-butyltin dichloride, min. 95%. MFCD00000518
SDP
Supplier:  Strem, An Ascensus Company 50113050G
Encompass_Preferred
CAS# 683-18-1. 50g. Di-n-butyltin dichloride, min. 95%. MFCD00000518. Molecular Weight: 303.83. Molecular Formula: (n-C4H9)2SnCl2. Color/form: white xtl. Strem# 50-1130. http://www.strem.com/catalog/v/50-1130/
Catalog No. 50-901-12586