Learn More
Cole-Parmer Worthington 4LDB Liquid Nitrogen Storage Dewar; 4 L, 10 Day Static Hold
Supplier: Cole-Parmer 0377351
Vacuum space is filled with technologically advanced insulation. Necktube core minimizes liquid nitrogen losses. Optional roller base lets you easily move larger capacity storage Dewars. Sturdy cast-aluminum frame has five wheels with ball-bearing swivel casters.