Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Detritylation Reagent, Honeywell Burdick & Jackson™
BioSyn™, suitable for use with DNA/RNA synthesis instruments, 3% Dichloroacetic Acid in Toluene (v/v)
$783.00 - $1820.00
Specifications
CAS | 79-43-6, 108-88-3 |
---|---|
CAS Max % | 3.00%, 97.00% |
Flash Point | 4°C.Toluene |
Physical Form | Liquid, clear |
Purity Grade Notes | Deblocking reagents |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
60-090-050
|
Honeywell Chemicals
BR6744X2.5GL |
4 x 2.5 L |
Case of 4 for $783.00
|
|
|||||
60-090-208
|
Honeywell Chemicals
BR67420 |
20 L |
Each for $1,820.00
|
|
|||||
Description
Burdick & Jackson® BioSyn® reagents & solvents for DNA/RNA synthesis
- Honeywell Burdick & Jackson® BioSyn® Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
- ideal for oligonucleotide synthesis regardless of the application
- All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination
Specifications
79-43-6, 108-88-3 | |
4°C.Toluene | |
Deblocking reagents |
3.00%, 97.00% | |
Liquid, clear |
Safety and Handling
Recommended Storage : Room Temperature
Spot an opportunity for improvement?
Product Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title