Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell Burdick & Jackson™ Detritylation Reagent, 3%Trichloroacetic Acid in Dichloromethane (withv)
BioSyn™, for DNA/RNA synthesis
$1790.00 - $4201.00
Specifications
CAS | 76-03-9 |
---|---|
Content And Storage | Room Temperature |
Density | 1.33 g/cm3 |
Packaging Type | Glass Bottle |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Packaging Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging Type | Price | Quantity & Availability | ||||
60-048-412
|
Honeywell Burdick & Jackson™
BR6052 |
4 x 2 L | Glass Bottle |
Case of 4 for $1,790.00
|
|
||||
60-048-413
|
Honeywell Burdick & Jackson™
BR6054 |
4 x 4 L | Glass Bottle |
Case of 4 for $4,201.00
|
|
||||
Description
Burdick & Jackson™ BioSyn™ reagents & solvents for DNA/RNA synthesis
- Honeywell Burdick & Jackson™ BioSyn™ Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
- ideal for oligonucleotide synthesis regardless of the application
- All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination
Specifications
76-03-9 | |
1.33 g/cm3 | |
Liquid |
Room Temperature | |
Glass Bottle |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title