Learn More
Labconco™ Detergent Dispenser Kit
Automates liquid detergent dispensing
Supplier: Labconco™ 4587500/EMD
Description
- Connect to any SteamScrubber™ or FlaskScrubber™ glassware scrubber
Specifications
Liquid Detergent Dispenser Kit | |
FlaskScrubber™, SteamScrubber™ glassware washer | |
10 lb. | |
Each |
To automate liquid detergent dispensing | |
Peristaltic pump, float switch, tubing, 1 gal. plastic container and installation instructions | |
4.5 kg |