Learn More
Biologix Technologies Inc Desktop Microtube Labels-White,
Supplier: Biologix Technologies Inc BLX038W
Circle top and a rectangle for the side. These Microtube labels fit all standard 1.5 – 2.0 ml Microcentrifuge tubes. Waterproof and freezer safe.2 rolls per pack500 labels per rollAll labels for use with DYMO® LabelWriter® printers