Learn More
Chemglass Life Sciences Desiccator, 150mm I.D., 2.4L, Small Top with 24/29 Stpk, 140mm Plate Dia, Complete
Supplier: Chemglass Life Sciences CG8272R150
Desiccator, 150mm I.D., 2.4L, Small Top with 24/29 Stpk, 140mm Plate Dia, Complete. This 2.4L Pyrex desiccator has a rugged 24/29 standard taper stopcock with tooled hose connections that serve as the vacuum inlet. It is designed so that its manipulation is not affected by the vacuum. To introduce vacuum into the desiccator, loosen the stopcock to disengage the PTFE plug from contact with the glass. A half turn from this position closes the desiccator. This design eliminates the need for grease. The tubulation O.D. is approximately 10mm. Approximate bowl volume capacity is 2,400mL. For replacement bowl see CG-8266-R150LO. For replacement stopcock see CG-8272-RSTPK. Please Note: Plate is not supplied and must be purchased seperately. WARNING: Do Not Heat Or Subject To Pressure. Rated 1-Atmosphere Vacuum.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.