Learn More
Perkin Elmer US LLC Desiccant Cartridge, Fitting, and Refill Kit for PGX-H2 Plus Generator
Supplier:  Perkin Elmer US LLC N9306064
Encompass_Preferred
This refill kit is for the PG-H2 Plus Pure Gas Hydrogen Generator. Pure gas hydrogen generators employ the newest membrane technology available for the safe production of pure hydrogen gas. This patented design is ideal for operation with gas analyzers, as fuel gas for flame tools, or as a source for pure hydrogen in plasma chambers and other isolated environments. Electrolytic membrane technology is preferred over alternative hydrogen generating techniques because it is clean, requires less maintenance and there is no need to store chemicals to maintain operation. The generators offer silent operation and require only deionized or distilled water with no caustic solutions which can affect the purity of the hydrogen.
Catalog No. 50-211-2911