Learn More
TARGETMOL CHEMICALS INC DESCHLOROCLOZAPINE 25MG
Supplier: TARGETMOL CHEMICALS INC T11078
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Deschloroclozapine a metabolite of Clozapine is a highly potent muscarinic DREADDs agonist. Deschloroclozapine binds to DREADD receptor subtypes hM3Dq and hM4Di with Ki of 6.3 and 4.2 nM respectively. purity: 99%