Learn More
Bachem Fmoc-b,b-diphenyl-Ala-OH
Supplier: Bachem 4030246.5000
Fmoc-b,b-diphenyl-Ala-OH, 201484-50-6, C30H25NO4, Mr 463.53, 5g. Fmoc-b-phenyl-Phe-OH, Fmoc-Dip-OH, Fmoc-3,3-diphenyl-L-alanine. Derivative for the introduction of Dip, a sterically hindered highly lipophilic phenylalanine substitute.