Learn More
Quality Biological Inc Water, DEPC Treated, 4 Liters
SDP
Supplier:  Quality Biological Inc 351068491
Encompass_Preferred
DEPC Treated Water is Ultra Pure and prepared by reverse osmosis, passed through fine carbon, de-ionized through two resin beds and serially filtered 1 ¿m positively charged membranes. ¿ The water is subsequently treated with 0.1% (v/v) DEPC. ¿ Used to reconstitute nucleic acids and/or proteins, and as a diluent or solvent for any molecular biology grade reagent. ¿ DEPC Treated Water may be usefully employed in RNA Applications (e.g., Preparations of an RNA probe, DNA Applications (e.g., Subcloning in pUC plasmids), and Protein Applications (e.g., Western Blotting). ¿ Storage Conditions: 15¿C to 30¿C ¿ Shipping Condition: ambient temperature ¿ Sterile Filtered & Deionized ¿ Endotoxin Tested < 0.03 EU/mL ¿ DNase, RNase, and Protease Tested ¿ Product and/or packaging contains recycled/renewable content ¿ Product and/or packaging is recyclable or reduces waste ¿ Product and/or packaging is safer to human and environmental health
Catalog No. 50-146-834