Learn More
Research Products International Corp DEPC, 100 ML
SDP
Supplier:  Research Products International Corp D43060100.0
Encompass_Preferred
Modification reagent for His and Tyr residues in proteins. Inhibitor of nucleases. DEPC treated water is used in handling of RNA in the laboratory to reduce the risk of RNA being degraded by RNases.Common Name: Diethyl PyrocarbonateCAS Number: 1609-47-8Molecular Weight: 162.1Chemical Formula: C6H10O5Solubility: AlcoholSTOR Temperature: 2-8CSTOR Conditions: RefrigeratorAppearance: Clear, Light YLW Liquid Identification: Pass Purity: 97.5 %Shipping Weight: 1 lbsShipping Dimensions: 5.11 x 5.11 x 5.11Product usually ships within 24 hours.Research or further manufacturing use only, not for food or drug use.
Catalog No. 50-223-0345