Learn More
Med Vet International Miltex Dental Scaler #12, Double-Ended, Round Handle
Supplier:  Med Vet International MIL70110
Encompass_Preferred
The Miltex 7-11 #12 dental scaler is made of OR grade German stainless steel. It has a round handle and is double-ended. This scaler can be used to remove supragingival calculus and deposits.
Catalog No. 50-264-3510
Explore available promotions