Learn More
Med Vet International Dental Curette Gracey 7/8
Supplier: Med Vet International DG7/8
Gracey curettes have only 1 cutting edge which is designed for deep subgingival calculus, root planing and curettage of the periodontal pocket.
This item is not returnable.
View return policy