Learn More
Cospheric LLC DENSITY MARKER BEADS KIT
Supplier: Cospheric LLC DMBKIT
Density Marker Beads Kit of 6 Densities, 2.5ml Vials, 20% concentration in Aqueous Solution. Colors by density are: Green-1.02g/cc, Orange-1.04g/cc, Violet-1.06g/cc, Dark Blue-1.08g/cc, Red-1.09g/cc, Med-Blue-1.13g/cc.