Learn More
United Scientific Supplies Density Cube Set
Set includes three different sized cubes of eight materials commonly used in density experiments
Supplier: United Scientific Supplies DCSET24
Description
- Cube set is composed of materials commonly used in density experiments
- Materials represented include acrylic, aluminum, brass, copper, nylon, polypropylene, PVC, and steel
- Reference table with density values included
- Each material is provided in three different sizes: 0.5, 1, and 1.5 in.
- 24 Cubes/Set
Specifications
24 Cubes/Set | |
Identifying Materials by Density | |
Density Cube Set | |
Acrylic, Aluminum, Brass, Copper, Nylon, Polypropylene, PVC, Steel |