Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Denka Seiken ™Agglutinating Sera, Salmonella H Slide Method
Denka Seiken™ Agglutinating Sera are packaged 2mL per vial.
Supplier: Thermo Scientific™ R679694
Description
Denka Seiken™ Agglutinating Sera are packaged 2mL per vial.
Specifications
Salmonella H-p (Slide Method) | |
2 mL/Vial | |
Salmonella |
Agglutinating Sera | |
2 mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction