Learn More
Thermo Scientific™ Denka Seiken™ Agglutinating Sera E. coli Polyvalent
Denka Seiken Agglutinating Sera, E. coli are packaged 2mL per vial.
Supplier:  Thermo Scientific™ R679054
 View more versions of this product
Catalog No. OXDK210548
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Description
Description
Denka Seiken Agglutinating Sera are packaged 2mL per vial.
  • Types O29, O143, O152, O164
TRUSTED_SUSTAINABILITY
Specifications
Specifications
E. coli Polyvalent 5
E. coli
Agglutinating Sera
2 mL/Vial