Learn More
Electron Microscopy Sciences Deluxe Saw Frame
SDP
Supplier:  Electron Microscopy Sciences 72009
Encompass_Preferred
A Swiss made saw frame lightweight and perfectly balanced. Its ergonomic wooden handle reduces fatigue and provides greater dexterity. Patented blade-lock design eases blade insertion and maintains perfect alignment which eliminates wobble and reduces blade loss during use. It measures 2 1/4" (70 mm) deep and accepts saw blades up to 1.2 mm diameter and 130 mm long. Used ideally with the Diamond Wire and Spiral Saw Blades.
Catalog No. 50-949-202