Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cellucap Deluxe Lab Coats
$255.00 - $450.00
Specifications
Material | Polypropylene |
---|---|
Color | White |
Closure Type | Snaps |
No. of Pockets | 3 |
For Use With (Application) | For splash protection from non-hazardous liquids and dry particulates |
Specifications
Polypropylene | |
Snaps | |
For splash protection from non-hazardous liquids and dry particulates | |
0.03 kg |
White | |
3 | |
0.06 lb. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title