Learn More
TECHNOTRADE INTERNATIONAL INC Deconex 11 ECONEX 11 Universal Cleaner 1kg
Supplier: TECHNOTRADE INTERNATIONAL INC 500100.00F10W
Deconex 11 UNIVERSAL is a mildly alkaline, liquid concentrate for the cleaning and decontamination of laboratory glassware. Can be universally used in different sectors of the chemical industry. Excellent substitute for chromic acid. 1kg