Learn More
VAI DECON-CYCLE™ II Germicidal Detergent
DECON-CYCLE™ II Germicidal Detergent is a low pH phenolic that is effective as hard non-porous surface disinfectant/detergent formulated for effective aseptic and controlled environment sanitation.
Supplier: VAI DCY2051Z
Includes: Lot-specific Certificate of Irradiation, Certificate of Analysis, LAL testing analysis and sterility report
Description
A low pH, dilutable, hospital detergent effective in 10 minutes at 20°C in hard water up to 400 ppm (calculated as CaCO3) in the presence of 5% blood serum. Recommended for use in pharmaceutical, biotechnology, medical device manufacturing, and in healthcare institutions to control cross-contamination hazards.
- Multi-phenolic formula designed to clean, disinfect, and deodorize any washable inanimate surface
- Filtered at 0.2 μm and gamma irradiated
- Strong enough to kill a broad spectrum of pathogenic bacteria, yet mild enough to have no harmful effect on disinfected surfaces
- Completely traceable and validated for sterility and shelf life
- Filled in an ISO 5 (Grade A/B, Former Class 100)
- Lot sterility tested according to current USP compendium
- Completely traceable from start to finish
Order Info
Double-bag packaging.
Compliance
Completely tested according to current USP compendium.
Specifications
Non-sterile | |
Kills broad spectrum of pathogenic Bacteria, pharmaceutical, biotechnology, medical device and health care industries | |
3.8 L |
SimpleMix | |
1 gal. |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.