Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VAI DECON-AHOL WFI Formula
DECON-AHOL WFI Formula is for disinfecting and sanitizing cleanrooms and controlled areas in health care institutions, biopharmaceutical, medical device, and diagnostic manufacturing facilities.
Supplier: VAI DECWFIBOT01
This item is not returnable.
View return policy
Description
- Low remaining residue
- Low in endotoxin levels
- For use on a multitude of surfaces
Specifications
Disinfectant | |
70% USP Isopropyl Alcohol, 30% USP Water for Injection | |
ASEPTI-CLEANSE Dispenser | |
946.4 mL |
Non-sterile | |
Bottle | |
32 oz. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction