Learn More
Perkin Elmer US LLC DC-2 Diamond Compression Cell
Supplier: Perkin Elmer US LLC L1272282
The DC-2 Diamond Compression Cell consists of two Hastelloy plates containing a pair of diamond windows that enable samples to be compressed to an ideal thickness for transmission measurements. This cell is compatible with the Microfocus Beam Condenser (L1272283/L1272284).