Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ DBI-SALA™ Retrax™ Shock Absorbing Lanyard
An alternative to elasticized lanyards. Uses a retraction technology like airport security lines. These devices retract and expand smoothly and consistently. The retraction always keeps the lanyard taut and close to the body, reducing trip fall hazards, snagging on equipment and dragging.
Supplier: 3M™ 7100240692
Description
Specifications
Yellow | |
Web | |
Variable/Retractable Shock Absorbing Lanyard | |
Snap Hook | |
User friendly self locking snap hooks at each end |
Web, Zinc-plated Steel | |
130 to 310 lb. | |
OSHA 1926.502, OSHA 1910.66 | |
6 ft. | |
1.8 m |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction