Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ DBI-SALA™ ExoFit NEX™ Construction Style Positioning Harness
3M™ DBI-SALA™ ExoFit NEX™ Construction Style Positioning Harness with ANSI, and OSHA safety compliance
Supplier: 3M™ 1113124
Description
- Hybrid shoulder, back and leg comfort padding
- Stand-up lightweight aluminum back D-ring
- Body belt/hip pad with lightweight aluminum side D-rings
- Web-locking quick-connect chest and leg straps
- Repel™ webbing
- Protective shoulder caps
- Built-in lanyard keepers
- Revolving vertical torso buckle
- Impact Indicator
- Integrated trauma straps
- Built-in reflective materials
- Protected labels, equipped with 3M™ Connected Safety ID (CSID)
Specifications
Duo-Lok™ Quick Connect | |
ANSI Z359.1, ANSI Z359.3, OSHA 1910.66, OSHA 1926.502 | |
Repel™ Polyester | |
190 kg | |
6.3 lb. |
Medium | |
6000 lb. | |
Construction Style Positioning Harness | |
420 lb. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction