Learn More
Cole-Parmer Casella CEL-350IS Individual Intrinsically Safe micro noise badge, with mounting clips
Supplier: Cole-Parmer 9893335
Easily position this noise badge on clothing or hard hats. It calculates all the popular noise dose indices and can store results for later and review. Smallonly 2.8" long, 1.7" wide and 2" tall-- and lightweight unit weighs less than 2.5 oz. Quick charge in less than 1.5 hours from completely empty.