Learn More
Steris™ Dart™ Daily Air Removal Test
Dart™ Daily Air Removal Test is used in monitoring healthcare steam sterilizers.
Supplier: Steris™ NB113
This item is not returnable.
View return policy
Description
Features:
- Disposable
- Preassembled Single Use test
- Functions as a AAMI Bowie Dick type test
- Light weight and compact
- Indicator strip can be kept as a permanent record
- Lot number and expiration date on each test
Benefits:
- Eliminates the need to construct test packs while providing consistent quality
- Non-uniform color change provides an immediate indication of inadequate air removal from the sterilizer chamber during the cycle
- Eliminates the need for bulky towels or wrappers
- Documentation of the efficacy of the sterilizer to record removed air from the pack
- Traceability and recognition of expired packs at time of use
- Used in monitoring healthcare steam sterilizers.
- Meets ANSI/AAMI/ISO 11140-4 requirements for Type 2 Air Removal Indicator Systems and operates at 270 to 274°F/132 to 134°C with an exposure time of 3.5 to 4.0 minutes.
- Easy to use.
- Users can eliminate waste associated with paper box products.
- Compact barrel design creates a barrier to air removal and steam penetration.
- Special record forms are included for convenience
User Process:
- Placed directly into an otherwise empty steam sterilizer chamber and does not require a retaining device.
- Optional tie is provided for suspension of the DART test.
- During processing, steam must displace the air through the blue heat sink and then within the restricted transparent chamber of the chemical indicator.
- Results are immediately visible through the clear vial.
- A uniform color change from yellow to black indicates that all the air was removed and replaced by steam.
- When the test is completed, simply snap off the window, slide out the chemical indicator, and record the results.
Specifications
ANSI/AAMI/ISO 11140-4 requirements for Type 2 Air Removal Indicator Systems | |
Disposable Preassembled Single Use Test | |
5 Pk./Cs. (10 Tests/Pk.) |