Learn More
Biocare Medical, LLC DAB Sparkle - 500 ml
SDP
Supplier:  Biocare Medical, LLC DS830M
Encompass_Preferred
This specially formulated ready-to-use enhancing solution will increase sensitivity and contrast of DAB stained tissues.
Catalog No. 50-828-24