Learn More
HITACHI HIGH TECH AMERICA INC Deuterium Lamp U1900/29XX/U-39XX
Supplier: HITACHI HIGH TECH AMERICA INC 2J11500
Replacement Deuterium lamp for use with U-1800, U-2800, U-2810, U-1900 / U2900 / U-2910 / U-3900 / U-3900H.