Learn More
Enzo Life Sciences D-erythro MAPP (5 mg). CAS: 143492-38-0
Supplier: Enzo Life Sciences BMLSL2210005
A cell-permeable and specific inhibitor of alkaline ceramidase, which can raise intracellular ceramide levels by blocking the hydrolysis of ceramide to sphingosine and free fatty acid. Enhances the apoptotic response to ionizing radiation by enhancing the ceramide signal. Alternative name: (1S,2R-D-erythro-2-N-Myristoylamino)-1-phenyl-1-propanol. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (25mg/ml) or 100% ethanol (25mg/ml). Long Term Storage: -20°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.