Learn More
Kamiya Biomedical D-Dimer Calibrator, 6x1mL/PK RX
SDP
Supplier:  Kamiya Biomedical KAI091C
Encompass_Preferred
K-ASSAY D-Dimer Calibrator. 6 x 1 mL (lyophilzed). For 6 point calibration. The K-ASSAY D-Dimer Calibrator is intended to be used for the multi-point calibration of the K-ASSAY D-Dimer assay (Cat. No. KAI-090) for the quantitative determination of cross-linked fibrin degradation products containing D-Dimer in human plasma or serum. FOR IN VITRO DIAGNOSTIC USE.
Catalog No. 50-465-256