Learn More
AdipoGen D-(+)-Camphoric acid (1 g)
SDP
Supplier:  AdipoGen AGCN20461G001
Encompass_Preferred
Chemical. CAS: 124-83-4. Formula: C10H16O4. MW: 200.2. Synthetic (originally isolated from Cinnamomum camphora). Chirality inducing agent. Useful tool as resolving agent to separate the isomers and purify racemic mixtures (e.g. DL-lysine) by fractionally crystallizing the product to obtain the pure isomer (e.g. L-lysine). Can also be used as building block. Insect repellent. Stimulator of osteoblast differentiation.
Catalog No. 50-205-3634