Learn More
ECM Bioscience Cytoskeletal Filament Labeling Immunocytochemistry Kit, ECM Biosciences
Supplier: ECM Bioscience CK7720
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The cytoskeletal filament kit can be used to label the major cytoskeletal elements in cells: actin filaments (Phalloidin:FITC), intermediate filaments (Vimentin), and microtubules (α-tubulin). The kit includes Phalloidin:FITC and Goat anti-mouse conjugated to DyLight 594 for for dual labeling experiments. Applications: ICC, IHC; Species Reactivity: Hu, Rt, Ms
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.