Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 16 Antibody (OTI2H8), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271085B
Description
Cytokeratin 16 Monoclonal specifically detects Cytokeratin 16 in Human, Rat samples. It is validated for Immunohistochemistry.Specifications
Cytokeratin 16 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
KRT16 | |
Full length recombinant protein of human Cytokeratin 16 (NP_005548) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Rat | |
IgG2a |
Immunohistochemistry | |
OTI2H8 | |
Immunohistochemistry | |
CK16, CK-16, cytokeratin 16, cytokeratin-16, FNEPPK, focal non-epidermolytic palmoplantar keratoderma, K16, K1CP, keratin 16, keratin, type I cytoskeletal 16, keratin-16, KRT16A, NEPPK | |
Mouse | |
Affinity Purified | |
Cytoskeleton Markers | |
3868.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction